MilliporeSigma™ Opticap™ XL2 Disposable Capsule Filters with Milligard™ Media: Single Layer Format
Specially designed for small flow applications or small flow rate liquid filtration
$704.47 - $736.90
Specifications
Autoclavable | Autoclavable |
---|---|
Material (Membrane) | MCE |
Sterility | Non-sterile |
Filtration Area | 1000 cm2 |
For Use With (Application) | Wine sample preparation; water filtration; and additives, aroma or adjuvants |
Catalog Number | Mfr. No. | Pore Size | Inlet Style | Outlet Style | Price | Quantity | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Pore Size | Inlet Style | Outlet Style | Price | Quantity | |||
KW03A02FF3
|
MilliporeSigma™
KW03A02FF3 |
0.2 μm | 0.75 in. (19mm) Sanitary Flange | 0.75 in. (19mm) Sanitary Flange |
Pack of 3 Each for $736.90
|
|
|||
KW03A02HH3
|
MilliporeSigma™
KW03A02HH3 |
0.2 μm | 0.56 in. (14mm) Hose Barb | 0.56 in. (14mm) Hose Barb |
Pack of 3 Each for $704.47
|
|
|||
KW03A02TT3
|
MilliporeSigma™
KW03A02TT3 |
0.2 μm | 1.5 in. (38mm) Sanitary Flange | 1.5 in. (38mm) Sanitary Flange |
Pack of 3 Each for $718.42
|
|
|||
KW06A02FF3
|
MilliporeSigma™
KW06A02FF3 |
0.5 μm | 0.75 in. (19mm) Sanitary Flange | 0.75 in. (19mm) Sanitary Flange |
Pack of 3 Each for $711.49
|
|
|||
KW06A02HH3
|
MilliporeSigma™
KW06A02HH3 |
0.5 μm | 0.56 in. (14mm) Hose Barb | 0.56 in. (14mm) Hose Barb |
Pack of 3 Each for $711.49
|
|
|||
KW06A02TT3
|
MilliporeSigma™
KW06A02TT3 |
0.5 μm | 1.5 in. (38mm) Sanitary Flange | 1.5 in. (38mm) Sanitary Flange |
Pack of 3 Each for $711.49
|
|
|||
KW19A02TT3
|
MilliporeSigma™
KW19A02TT3 |
1.2 μm | 1.5 in. (38mm) Sanitary Flange | 1.5 in. (38mm) Sanitary Flange |
Pack of 3 Each for $718.42
|
|
|||
KW19A02HH3
|
MilliporeSigma™
KW19A02HH3 |
1.2 μm | 0.56 in. (14mm) Hose Barb | 0.56 in. (14mm) Hose Barb |
Pack of 3 Each for $704.47
|
|
|||
KW19A02FF3
|
MilliporeSigma™
KW19A02FF3 |
1.2 μm | 0.75 in. (19mm) Sanitary Flange | 0.75 in. (19mm) Sanitary Flange |
Pack of 3 Each for $704.47
|
|
|||
Description
- Integrated pleat pack of Milligard filter media provides high retention efficiency for the removal of particulate and microbiological contaminant
- Provide ease of use and convenience by eliminating the need for cartridge housing manipulations
- High spot vent and low spot drain ensure easy venting/ draining
- Unique capsule design contributes to minimal hold-up volume
Not in-line steam sterilizable
Wine sample preparation, water filtration, and additives, aroma or adjuvants
Order Info
Packaged in double easy-open bag. Manufactured in accordance with a Quality Management System approved by an accredited registering body to an ISO™ 9000 Quality Systems Standard and are shipped with a Certificate of Quality.
Specifications
Autoclavable | |
Non-sterile | |
Wine sample preparation; water filtration; and additives, aroma or adjuvants | |
5.6 in. |
MCE | |
1000 cm2 | |
Single Layer | |
14.2 cm |