MilliporeSigma™ ReBlot Plus Kit and Reagents
Blots can be stripped of antibodies in approximately 15 minutes at room temperature
Supplier: MilliporeSigma™ 2500
Description
Western blotting is a commonly used technique for studying protein function and localization. Typically, protein samples are electrophoresed on SDS-PAGE and transferred to a membrane such as nitrocellulose or nylon, where they are probed with specific antibodies. Unlike nucleic acid-based technologies, which allow reuse of Southern and Northern blots, it has been difficult to reuse Western blots.
- No pungent-smelling β-mercaptoethanol is contained in the Antibody Stripping Solution
- Antibody stripping is done at room temperature—no heating of blots is required
- Blots may be reused in 25 min.
Specifications
Western Blot Kit | |
Ea. | |
4°C |
Western Blot | |
50mL Mild Antibody stripping solution 10X, 50mL strong Antibody stripping solution 10X |