missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
MilliporeSigma™ ReBlot Plus Kit and Reagents
Blots can be stripped of antibodies in approximately 15 minutes at room temperature
$213.00 - $381.96
Specifications
For Use With (Application) | Western Blot |
---|
Includes: Mild Antibody Stripping Solution (10X), 50mL; Strong Antibody Stripping Solution (10X), 50mL
Catalog Number | Mfr. No. | Product Type | Quantity | Kit Contents | Content And Storage | Price | Quantity | ||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Product Type | Quantity | Kit Contents | Content And Storage | Price | Quantity | ||
2500MI
|
MilliporeSigma™
2500 |
Western Blot Kit | Ea. | 50mL Mild Antibody stripping solution 10X, 50mL strong Antibody stripping solution 10X | 4°C |
Each for $381.96
|
|
||
2502MI
|
MilliporeSigma™
2502 |
Western Blot Stripping Buffer | 50 mL | 25mL Mild Antibody stripping solution 10X | 2°C to 8°C |
Each for $213.00
|
|
||
Description
Western blotting is a commonly used technique for studying protein function and localization. Typically, protein samples are electrophoresed on SDS-PAGE and transferred to a membrane such as nitrocellulose or nylon, where they are probed with specific antibodies. Unlike nucleic acid-based technologies, which allow reuse of Southern and Northern blots, it has been difficult to reuse Western blots.
- No pungent-smelling β-mercaptoethanol is contained in the Antibody Stripping Solution
- Antibody stripping is done at room temperature—no heating of blots is required
- Blots may be reused in 25 min.
Specifications
Western Blot |