MilliporeSigma™ Silica Gel 60 F254 Coated Aluminum-Backed TLC Sheets
Allows analyzation of any substance when combined with suitable mobile phase
Supplier:  MilliporeSigma™ 1055490001
 View more versions of this product
Catalog No. M1055490001
Explore available promotions
Description
Description
  • Combination of unmodified silica gel 60 and polymeric binder for hard and adherent surface, resistant to crack and damage
  • Dense plate coating ensuring narrow bands and maximum sample separation efficiency
  • Layer thickness of 200μm with mean particle size of 10 to 12μm
  • Can be cut with scissors for individual separation requirements
  • Green fluorescencing inorganic florescent indications for UV detection of colorless substance
  • Available in broad range of sizes
  • Disposable
  • Fibre case packaging
  • In-process control in drug synthesis
  • Identity and stability testing of drugs
  • Quality control of pharmaceuticals, food and environmental samples
  • Residue analysis in food and environmental samples
Specifications
Specifications
20 to 200 min.
10 to 12 μm
20/Pk.
Single-use
In-Process Control in Drug Synthesis, Identity and Stability Testing of Drugs, Quality Control of Pharmaceuticals, Food and Environmental Samples, Residue Analysis in Food and Environmental Samples
5 to 20 μm
Silica Gel 60 F254
200 μm
7.5 cm
0.0011 in.
Aluminum
60 Å
Classical Silica TLC Plate
254 nm
10.0
0.8 mL/g
520 m2/g
5 x 7.5 cm
5 cm
30 μm
Product Suggestions
Product Suggestions