MilliporeSigma™ Silica Gel 60 F254 Coated Aluminum-Backed TLC Sheets
Allows analyzation of any substance when combined with suitable mobile phase
$95.71 - $1274.39
Specifications
Backing Material Aluminum
Pore Size 60 Å
Excitation Wavelength 254 nm
Thickness (Metric) 200 μm
View More Specs
Products 8
Catalog Number Mfr. No. Dimensions (L x W) Quantity Type Price Quantity  
Catalog Number Mfr. No. Dimensions (L x W) Quantity Type Price Quantity  
M1055340001
View Documents
MilliporeSigma™
1055340001
5 x 20 cm 100/Pk. Classical Silica TLC Plate
Each for $516.84
M1055480001
View Documents
MilliporeSigma™
1055480001
20 x 20 cm 25/Pk. Silica HPTLC Plate
Each for $724.11
M1055490001
View Documents
MilliporeSigma™
1055490001
5 x 7.5 cm 20/Pk. Classical Silica TLC Plate
Each for $95.71
M1055590001
View Documents
MilliporeSigma™
1055590001
20 x 20 cm 20/Pk. RP-modified Silica TLC Plate
Each for $1,274.39
M1055540001
View Documents Promotion Details
MilliporeSigma™
1055540001
20 x 20 cm 25/Pk. Classical Silica TLC Plate
Each for $346.21
M1055600001
View Documents
MilliporeSigma™
1055600001
5 x 7.5 cm 20/Pk. RP-modified Silica TLC Plate
Each for $260.63
M1055830001
View Documents
MilliporeSigma™
1055830001
20 x 20 cm 25/Pk. Concentrating Zone TLC Plate
Each for $629.53
M1168340001
View Documents
MilliporeSigma™
1168340001
5 x 10 cm 50/Pk. Classical Silica TLC Plate
Each for $526.45
Description
Description
  • Combination of unmodified silica gel 60 and polymeric binder for hard and adherent surface, resistant to crack and damage
  • Dense plate coating ensuring narrow bands and maximum sample separation efficiency
  • Layer thickness of 200μm with mean particle size of 10 to 12μm
  • Can be cut with scissors for individual separation requirements
  • Green fluorescencing inorganic florescent indications for UV detection of colorless substance
  • Available in broad range of sizes
  • Disposable
  • Fibre case packaging
  • In-process control in drug synthesis
  • Identity and stability testing of drugs
  • Quality control of pharmaceuticals, food and environmental samples
  • Residue analysis in food and environmental samples
Specifications
Specifications
Aluminum
60 Å
254 nm
200 μm
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'