MilliporeSigma™ TLC Silica Gel 60 Aluminum Sheets
For thin layer chromatography applications.
Supplier:  MilliporeSigma™ 1055820001
 View more versions of this product
Catalog No. M1055820001
Explore available promotions
Description
Description
Our classical silica TLC plates are based on a combination of our proven silica gel 60 and the addition of a unique polymeric binder resulting in a very adherent and hard surface that will not crack or blister and even allow writing with a pensil on the surface without risk to damage the layer.
Our concentrating zone plates are based on different adsorption properties of two adsorbents. The first is a large pore concentrating adsorbent where the samples are applied; the second is a selective layer for separation. Regardless of the spots’ shape, size, or position, the sample always concentrates as a narrow band where the two adsorbents overlap and where the separation starts. These plates allow users to quickly and easily apply any kind of sample, even large volumes of diluted samples.
Specifications
Specifications
Aluminum
25/Pk.
Quick and Easy Sample AppliCatIon From Small to Large Volumes of Diluted Samples, Highly Facilitated Sample Loading, Better Resolution Due to Focused Bands
20 x 2.5 cm
200 μm
20 cm
60 Å
Concentrating Zone TLC Plate
7.87 x 0.98 in.
Silica Gel 60 with Concentrating Zone
20 x 20 cm
20 cm