MilliporeSigma™ TLC Silica Gel 60 Aluminum Sheets
For thin layer chromatography applications.
$396.89 - $574.79
Specifications
Backing Material Aluminum
Pore Size 60 Å
Quantity 25/Pk.
Thickness (Metric) 200 μm
Dimensions (L x W) 20 x 20 cm
View More Specs
Products 2
Catalog Number Mfr. No. Sorbent Price Quantity  
Catalog Number Mfr. No. Sorbent Price Quantity  
M1055530001
View Documents
MilliporeSigma™
1055530001
Silica Gel 60
Each for $396.89
M1055820001
View Documents
MilliporeSigma™
1055820001
Silica Gel 60 with Concentrating Zone
Each for $574.79
Description
Description
Our classical silica TLC plates are based on a combination of our proven silica gel 60 and the addition of a unique polymeric binder resulting in a very adherent and hard surface that will not crack or blister and even allow writing with a pensil on the surface without risk to damage the layer.
Our concentrating zone plates are based on different adsorption properties of two adsorbents. The first is a large pore concentrating adsorbent where the samples are applied; the second is a selective layer for separation. Regardless of the spots’ shape, size, or position, the sample always concentrates as a narrow band where the two adsorbents overlap and where the separation starts. These plates allow users to quickly and easily apply any kind of sample, even large volumes of diluted samples.
Specifications
Specifications
Aluminum
25/Pk.
20 x 20 cm
20 cm
60 Å
200 μm
20 cm
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'