missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Emodin, Thermo Scientific Chemicals

A protein tyrosine kinase inhibitor

Supplier:  Thermo Scientific Chemicals J61600MD

 View more versions of this product

Catalog No. AAJ61600MD


Explore available promotions

Description

Description

A protein tyrosine kinase inhibitor

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Emodin
Orange
MFCD00001207
1888141
emodin, schuttgelb, emodol, frangula emodin, rheum emodin, frangulic acid, 3-methyl-1,6,8-trihydroxyanthraquinone, archin, persian berry lake, 1,3,8-trihydroxy-6-methyl-9,10-anthraquinone
CC1=CC(=C2C(=C1)C(=O)C3=CC(=CC(=C3C2=O)O)O)O
270.24
CHEBI:42223
Powder
518-82-1
C15H10O5
250 mg
14,3561
RHMXXJGYXNZAPX-UHFFFAOYSA-N
1,3,8-trihydroxy-6-methylanthracene-9,10-dione
3220
270.24

Chemical Identifiers

518-82-1
270.24
RHMXXJGYXNZAPX-UHFFFAOYSA-N
3220
1,3,8-trihydroxy-6-methylanthracene-9,10-dione
C15H10O5
MFCD00001207
emodin, schuttgelb, emodol, frangula emodin, rheum emodin, frangulic acid, 3-methyl-1,6,8-trihydroxyanthraquinone, archin, persian berry lake, 1,3,8-trihydroxy-6-methyl-9,10-anthraquinone
CHEBI:42223
CC1=CC(=C2C(=C1)C(=O)C3=CC(=CC(=C3C2=O)O)O)O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 208-258-8

RTECSNumber : CB7920600

TSCA : No

Recommended Storage : Keep cold

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only