Learn More
Emodin, Thermo Scientific Chemicals
A protein tyrosine kinase inhibitor
$277.19 - $1354.49
Chemical Identifiers
CAS | 518-82-1 |
---|---|
Molecular Formula | C15H10O5 |
Molecular Weight (g/mol) | 270.24 |
MDL Number | MFCD00001207 |
InChI Key | RHMXXJGYXNZAPX-UHFFFAOYSA-N |
Synonym | emodin, schuttgelb, emodol, frangula emodin, rheum emodin, frangulic acid, 3-methyl-1,6,8-trihydroxyanthraquinone, archin, persian berry lake, 1,3,8-trihydroxy-6-methyl-9,10-anthraquinone |
PubChem CID | 3220 |
ChEBI | CHEBI:42223 |
IUPAC Name | 1,3,8-trihydroxy-6-methylanthracene-9,10-dione |
SMILES | CC1=CC(=C2C(=C1)C(=O)C3=CC(=CC(=C3C2=O)O)O)O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAJ61600MC
|
Thermo Scientific Chemicals
J61600MC |
100 mg |
Each for $277.19
|
|
|||||
AAJ61600MD
|
Thermo Scientific Chemicals
J61600MD |
250 mg |
Each for $551.01
|
|
|||||
AAJ6160003
|
Thermo Scientific Chemicals
J6160003 |
1 g |
Each for $1,354.49
|
|
|||||
Description
A protein tyrosine kinase inhibitor
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
518-82-1 | |
270.24 | |
RHMXXJGYXNZAPX-UHFFFAOYSA-N | |
3220 | |
1,3,8-trihydroxy-6-methylanthracene-9,10-dione |
C15H10O5 | |
MFCD00001207 | |
emodin, schuttgelb, emodol, frangula emodin, rheum emodin, frangulic acid, 3-methyl-1,6,8-trihydroxyanthraquinone, archin, persian berry lake, 1,3,8-trihydroxy-6-methyl-9,10-anthraquinone | |
CHEBI:42223 | |
CC1=CC(=C2C(=C1)C(=O)C3=CC(=CC(=C3C2=O)O)O)O |
Specifications
518-82-1 | |
C15H10O5 | |
100 mg | |
14,3561 | |
RHMXXJGYXNZAPX-UHFFFAOYSA-N | |
1,3,8-trihydroxy-6-methylanthracene-9,10-dione | |
3220 | |
270.24 | |
Emodin |
Orange | |
MFCD00001207 | |
1888141 | |
emodin, schuttgelb, emodol, frangula emodin, rheum emodin, frangulic acid, 3-methyl-1,6,8-trihydroxyanthraquinone, archin, persian berry lake, 1,3,8-trihydroxy-6-methyl-9,10-anthraquinone | |
CC1=CC(=C2C(=C1)C(=O)C3=CC(=CC(=C3C2=O)O)O)O | |
270.24 | |
CHEBI:42223 | |
Powder |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 208-258-8
RTECSNumber : CB7920600
TSCA : No
Recommended Storage : Keep cold
RUO – Research Use Only