Ansell ENCORE™ Perry Style 42™ PF Surgical Gloves
Latex, powder free surgical gloves feature a smooth finish and optimized thickness. Ideal for brief, low-risk sterile procedures inside or outside of the operating room.
Supplier:  Ansell 5711102PF
 View more versions of this product
Catalog No. 19-501-153
Explore available promotions
Description
Description
  • Natural rubber latex
  • Made with proprietary anatomic formers for comfort and to help reduce hand fatigue
  • Polymer coated for fast and easy donning
  • Optimized glove thickness for the best balance of protection and tactile sensitivity
  • Recommended for:
    • Brief, low-risk examination procedures
    • Sterile procedures in the O.R.
    • Use as an outer glove when double gloving
    • Sterile procedures outside of the O.R.
Specifications
Specifications
ASTM D3577
0.65
No Texture
Latex
Sterile
5711102PF
EN 420:2003 + A1:2009
Natural
No
6.5
ISO 13485, ISO 9001
ENCORE™ Perry Style 42™ PF