Ansell ENCORE™ Perry Style 42™ PF Surgical Gloves
Latex, powder free surgical gloves feature a smooth finish and optimized thickness. Ideal for brief, low-risk sterile procedures inside or outside of the operating room.
$253.94 - $253.94
Specifications
Certifications/Compliance ASTM D3577
Certifications/Compliance (Europe) EN 420:2003 + A1:2009
AQL 0.65
Color Natural
Exterior Finish No Texture
View More Specs
Products 2
Catalog Number Mfr. No. Size Price Quantity  
Catalog Number Mfr. No. Size Price Quantity  
19-501-153
View Documents
Ansell
5711102PF
6.5 N/A
19-501-157
View Documents
Ansell
5711106PF
8.5 N/A
Description
Description
  • Natural rubber latex
  • Made with proprietary anatomic formers for comfort and to help reduce hand fatigue
  • Polymer coated for fast and easy donning
  • Optimized glove thickness for the best balance of protection and tactile sensitivity
  • Recommended for:
    • Brief, low-risk examination procedures
    • Sterile procedures in the O.R.
    • Use as an outer glove when double gloving
    • Sterile procedures outside of the O.R.
Specifications
Specifications
ASTM D3577
0.65
No Texture
Latex
ISO 13485, ISO 9001
EN 420:2003 + A1:2009
Natural
No
Sterile
ENCORE™ Perry Style 42™ PF
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'