ENG Scientific RBC Diluting Fluid (Gower's Solution)
RBC Diluting Fluid (Gower's Solution) is used for diluting RBC's. Contains Sodium sulfate and Glacial acetic acid. Settling time is between 1 to 3 minutes.
Supplier: ENG Scientific 5603
Description
- Contains: Sodium sulfate, Glacial acetic acid
Specifications
1 x 100 mL | |
For in-vitro diagnostic use only | |
1 to 3 min. | |
Bottles |
For in vitro diagnostic use only.