missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
ENG Scientific RBC Diluting Fluid (Gower's Solution)
RBC Diluting Fluid (Gower's Solution) is used for diluting RBC's. Contains Sodium sulfate and Glacial acetic acid. Settling time is between 1 to 3 minutes.
$492.65 - $492.65
Specifications
For Use With (Application) | For in-vitro diagnostic use only |
---|---|
Staining Time | 1 to 3 min. |
Packaging | Bottles |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
ES5603
|
ENG Scientific
5603 |
1 x 100 mL |
Each for $492.65
|
|
|||||
Description
- Contains: Sodium sulfate, Glacial acetic acid
Specifications
For in-vitro diagnostic use only | |
1 to 3 min. | |
Bottles |
For in vitro diagnostic use only.