missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Enoxacin Sesquihydrate 98.0+%, TCI America™

Supplier:  TCI America E076225G

 View more versions of this product

Catalog No. E076225G


Explore available promotions

Description

Description

Specifications

Specifications

Enoxacin Sesquihydrate
226°C
C30H36F2N8O7
1-Ethyl-6-fluoro-1,4-dihydro-4-oxo-7-(1-piperazinyl)-1,8-naphthyridine-3-carboxylic Acid
CCN1C=C(C(=O)C2=CC(=C(N=C21)N3CCNCC3)F)C(=O)O.CCN1C=C(C(=O)C2=CC(=C(N=C21)N3CCNCC3)F)C(=O)O.O
658.664
320.32
Crystalline Powder
84294-96-2
White-Yellow
25 g
KIJBMNCGVILNLN-UHFFFAOYSA-N
1-ethyl-6-fluoro-4-oxo-7-piperazin-1-yl-1,8-naphthyridine-3-carboxylic acid;hydrate
91659135
≥98.0% (HPLC,T)

Chemical Identifiers

84294-96-2
658.664
1-Ethyl-6-fluoro-1,4-dihydro-4-oxo-7-(1-piperazinyl)-1,8-naphthyridine-3-carboxylic Acid
1-ethyl-6-fluoro-4-oxo-7-piperazin-1-yl-1,8-naphthyridine-3-carboxylic acid;hydrate
C30H36F2N8O7
KIJBMNCGVILNLN-UHFFFAOYSA-N
91659135
CCN1C=C(C(=O)C2=CC(=C(N=C21)N3CCNCC3)F)C(=O)O.CCN1C=C(C(=O)C2=CC(=C(N=C21)N3CCNCC3)F)C(=O)O.O
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents

Promotions

Promotions