missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Enoxacin Sesquihydrate 98.0+%, TCI America™
$233.92 - $691.81
Chemical Identifiers
CAS | 84294-96-2 |
---|---|
Molecular Formula | C30H36F2N8O7 |
Molecular Weight (g/mol) | 658.664 |
InChI Key | KIJBMNCGVILNLN-UHFFFAOYSA-N |
Synonym | 1-Ethyl-6-fluoro-1,4-dihydro-4-oxo-7-(1-piperazinyl)-1,8-naphthyridine-3-carboxylic Acid |
PubChem CID | 91659135 |
IUPAC Name | 1-ethyl-6-fluoro-4-oxo-7-piperazin-1-yl-1,8-naphthyridine-3-carboxylic acid;hydrate |
SMILES | CCN1C=C(C(=O)C2=CC(=C(N=C21)N3CCNCC3)F)C(=O)O.CCN1C=C(C(=O)C2=CC(=C(N=C21)N3CCNCC3)F)C(=O)O.O |
Chemical Identifiers
84294-96-2 | |
658.664 | |
1-Ethyl-6-fluoro-1,4-dihydro-4-oxo-7-(1-piperazinyl)-1,8-naphthyridine-3-carboxylic Acid | |
1-ethyl-6-fluoro-4-oxo-7-piperazin-1-yl-1,8-naphthyridine-3-carboxylic acid;hydrate |
C30H36F2N8O7 | |
KIJBMNCGVILNLN-UHFFFAOYSA-N | |
91659135 | |
CCN1C=C(C(=O)C2=CC(=C(N=C21)N3CCNCC3)F)C(=O)O.CCN1C=C(C(=O)C2=CC(=C(N=C21)N3CCNCC3)F)C(=O)O.O |
Specifications
84294-96-2 | |
White-Yellow | |
5 g | |
KIJBMNCGVILNLN-UHFFFAOYSA-N | |
1-ethyl-6-fluoro-4-oxo-7-piperazin-1-yl-1,8-naphthyridine-3-carboxylic acid;hydrate | |
91659135 | |
≥98.0% (HPLC,T) | |
Enoxacin Sesquihydrate |
226°C | |
C30H36F2N8O7 | |
1-Ethyl-6-fluoro-1,4-dihydro-4-oxo-7-(1-piperazinyl)-1,8-naphthyridine-3-carboxylic Acid | |
CCN1C=C(C(=O)C2=CC(=C(N=C21)N3CCNCC3)F)C(=O)O.CCN1C=C(C(=O)C2=CC(=C(N=C21)N3CCNCC3)F)C(=O)O.O | |
658.664 | |
320.32 | |
Crystalline Powder |
Safety and Handling
TSCA : No