Eppendorf™ Rotor for Centrifuges 5804 R and 5810 R
Eppendorf Rotor for Centrifuges 5804 R and 5810 R-precision meets power. Customize your centrifugation with varied capacities and speeds. Accessorize with versatile buckets and adaptors for efficient sample processing. Ideal for high-performance applications, ensuring accuracy and reproducibility.
Supplier: Eppendorf™ 022637100
Description
- Precision-engineered for optimal sample separation.
- All fixed-angle rotors, rotor lids, buckets, caps and adapters can be autoclaved (121°C, 20 min.).
- Accessories (adaptors and buckets) further customize your centrifuge setup.
- Aerosol-tight options available.
Order Info
Can also accept 0.5 and 0.4/0.25 mL tubes when equipped with accessory adapters (05-406-10 and 05-406-25, respectively; order separately)
Specifications
Aluminum | |
30 x 1.5/2 mL | |
14,000 rpm | |
Aerosol-tight lid |
Fixed Angle Rotor | |
20,800 x g | |
5804/5810 Centrifuge | |
Autoclavable |