Eppendorf™ Rotor for Centrifuges 5804 R and 5810 R
Eppendorf Rotor for Centrifuges 5804 R and 5810 R-precision meets power. Customize your centrifugation with varied capacities and speeds. Accessorize with versatile buckets and adaptors for efficient sample processing. Ideal for high-performance applications, ensuring accuracy and reproducibility.
Supplier:  Eppendorf™ 022638564
 View more versions of this product
Catalog No. 05-40-104
Explore available promotions
Description
Description
  • Precision-engineered for optimal sample separation.
  • All fixed-angle rotors, rotor lids, buckets, caps and adapters can be autoclaved (121°C, 20 min.).
  • Accessories (adaptors and buckets) further customize your centrifuge setup.
  • Aerosol-tight options available.
Specifications
Specifications
A-2-DWP, for Deepwell plates - Includes buckets (x2)
2 x 5 MTP, 2 x 2 DWP
3,700 rpm
Includes buckets (x2)
Rotor
2,250 x g
Centrifuges 5804 R and 5810 R
Autoclavable