Eppendorf™ Rotor for Centrifuges 5804 R and 5810 R
Eppendorf Rotor for Centrifuges 5804 R and 5810 R-precision meets power. Customize your centrifugation with varied capacities and speeds. Accessorize with versatile buckets and adaptors for efficient sample processing. Ideal for high-performance applications, ensuring accuracy and reproducibility.
Supplier:  Eppendorf™ 5820765003
 View more versions of this product
Catalog No. 5820765003
Explore available promotions
Description
Description
  • Precision-engineered for optimal sample separation.
  • All fixed-angle rotors, rotor lids, buckets, caps and adapters can be autoclaved (121°C, 20 min.).
  • Accessories (adaptors and buckets) further customize your centrifuge setup.
  • Aerosol-tight options available.
Specifications
Specifications
FA-45-20-17, Aerosol-tight - Includes rotor lid
20 x 5.0 mL
13,100 rpm
Rotor lid
Rotor
20,913 x g
Centrifuges 5804 R and 5810 R