Eppendorf™ epT.I.P.S.™ Singles - Pipette Tips
Eppendorf epT.I.P.S.™ Singles high-precision pipette tips offer unparalleled precision in 'Biopur™' and 'Sterile' grades, featuring individually wrapped tips for maximum sterility, ideal for sensitive and critical lab applications where contamination control is paramount.
Supplier:  Eppendorf™ 022491156
 View more versions of this product
Catalog No. 05-403-07
Explore available promotions
Description
Description
  • Eppendorf pipette tip, individually wrapped
  • Available in Biopur and Sterile purity grades
  • For use in medical, pharmaceutical, food industry, molecular biology and cell technology
  • Batch number and expiration date on each blister package
  • Each production lot inspected continuously by an independent laboratory
  • Lot-specific certificates available
  • ACT™-labeled by non-profit My Green Lab™ to help you make an informed product choice when it comes to sustainability
Specifications
Specifications
50 to 1000 μL
Sterile
Autoclavable
71 mm
Biopur™ (Sterile)
Individually Wrapped
100 Tips, Individually Packed
Blue
Air-cushion System
Biopur
Fine
Non-filtered
Colorless
2.79 in.
ISO 9001
Eppendorf Pipettes, Non-Eppendorf Pipettes
Extended Length, Finely Tapered, Universal
Pipette Tips
Single-use
Yes
Polypropylene