Eppendorf™ Centrifuge Bottles
Wide mouth plastic centrifuge bottles.
Supplier:  Eppendorf™ 022638653
 View more versions of this product
Catalog No. 36-100-7281
Explore available promotions
Description
Description
For use with Eppendorf Centrifuges 5804/5804R, 5810/5810R.
Specifications
Specifications
13 oz., 400mL
13 oz.
Plastic
3250 x g
1 x 400mL Adapter for super capacity 4 x 400mL rotor A-4-81
Screw Cap
400 mL
Screw cap
Autoclavable
Wide
Polypropylene