Eppendorf™ Combitips Advanced™ Biopur™ Pipette Tips
Meet the increased requirements of a modern laboratory with these pipetter tips. Eppendorf™ Combitips Advanced™ Biopur™ Pipetter Tips make working with radioactive, aggressive or toxic materials safer. Hermetically sealed piston prevents aerosol contamination.
Supplier:  Eppendorf™ 0030089642
 View more versions of this product
Catalog No. 13-683-721
Explore available promotions
Description
Description
Meet the increased requirements of a modern laboratory with these pipetter tips. Eppendorf™ Combitips Advanced™ Biopur Pipetter Tips make working with radioactive, aggressive or toxic materials safer. Hermetically sealed piston prevents aerosol contamination.
Combitips advanced are the ideal components for all Multipettes™. The new individual color coding facilitates quick identification of the desired tip.
  • Slip agent free
  • Color-coded tip size recognition
  • Funnel geometry from cylinder to rim, prevents damage to gloves and ensures comfortable handling
  • Function according to the positive displacement principle
  • Elongated design for complete emptying of all common tubes
  • Optimized graduations for visibility of liquid volume
  • Automatic Combitip recognition and volume calculation
  • 3D design on the upper rim for easy recognition
  • Maximum range offered by nine volume sizes (0.1 to 50 mL)
  • Dust free storage
Specifications
Specifications
1 mL
Colorless
Biopur™ (Sterile)
Extended Length, Positive Displacement
Pipette Tips
Single-use
Yes
Polypropylene
Sterile
Not Autoclavable
Individually Wrapped
100 Pk., Individually Blister-wrapped
Yellow
Eppendorf Multipette
CombiTips Advanced
Sterile & PCR Ready