Eppendorf™ Heat Sealer Film and Foils
Create 100% hermetic seal, eliminate evaporation, and prevent cross-contamination
$289.76 - $486.86
Catalog Number | Mfr. No. | For Use With (Application) | Product Type | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | For Use With (Application) | Product Type | Price | Quantity | ||||
E0030127870
|
Eppendorf™
0030127870 |
High-strength adhesive that forms tight seals to minimize sample evaporation | Storage Film |
Pack of 100 Each for $480.39
|
|
||||
E0030127854
|
Eppendorf™
0030127854 |
Heat sealing PCR foil for sealing plates, especially recommended for low reaction volumes, best protection against evaporation | PCR Foil |
Pack of 100 Each for $289.76
|
|
||||
E0030127838
|
Eppendorf™
0030127838 |
Hermetic sealing of PCR plates, especially recommended for low reaction volumes, best protection against evaporation | PCR Film |
Pack of 100 Each for $486.86
|
|
||||
Description
- Requires Eppendorf heat sealer (Cat. No. E5390 000 016)
- Transparent heat sealing film allows sample monitoring in the plates
- Aluminum coated, heat-sealing foils are available in peel-off or pierceable versions
- Foil stripper tool removes heat-sealed film and foils completely
- Certified free of DNA, DNase, RNase, and PCR inhibitors
- Autoclavable
Specifications
Autoclavable | |
Yes |