Eppendorf™ HeatSealer S200 Accessory, Plate Adapters
For use with HeatSealer S200
Supplier:  Eppendorf™ 5392070020
 View more versions of this product
Catalog No. 05-412-616
Explore available promotions
Description
Description
  • Hermetic sealing of multiwell plates
  • Eliminates evaporation in PCR, reducing cross contamination
  • Seals 96- and 384-well plates
  • Integrated thermostat prevents overheating
  • Improved mechanics for even easier sealing
  • PCR Plate Adapter included
Specifications
Specifications
Low Profile Adapter
350 W
HeatSealer S200
125°C to 200°C in 1°C increments