Eppendorf™ MiniSpin™ / MiniSpin™ Plus - Mini Centrifuge
Eppendorf MiniSpin™ and MiniSpin™ plus centrifuges are powerful, entry-level, easy-to-use centrifuges that are small enough to allow individual workstations to be equipped with their own. Key parts are made of metal for maximum safety, and the MiniSpin™ plus is fast enough for molecular biology separations.
Supplier:  Eppendorf™ 022620207
 View more versions of this product
Catalog No. 05-401-07
Explore available promotions
Description
Description
  • Extremely compact footprint
  • Acceleration and deceleration time <13 sec.
  • User-friendly digital display for time and speed
  • Metal rotor bowl
  • Separate short-spin key for fast and comfortable centrifugation
  • Optimal ventilation flow reduces heating and protects temperature-sensitive samples
  • Soft-touch lid closure for ergonomic lid locking
  • Automatic lid opening at the end of the run to prevent sample warming and to allow easy access to samples
Special features for MiniSpin plus:
  • Automatic rpm/rcf conversion
  • Extended time to 99 min or continuous centrifugation
Certifications
CE, CSA
Compliance
Meet international safety standards for centrifuges (IEC 1010-2-020)
Specifications
Specifications
12 x 1.5/2.0 mL Tubes (w/ inc. rotor)
14,100 x g (w/ incl. rotor)
<60 dB(A)
12-place Rotor, Cover
MiniSpin™ plus, 120 V - Includes: Rotor F-45-12-11
22.5 x 23.0 x 13.0 cm (8.86 x 9.06 x 5.12 in.)
14,000 to 14,999 x g
Yes
99 min.
120 V
14,500 rpm (w/ incl. rotor)
Color Graphic Display
Meet international safety standards for centrifuges (IEC 1010-2-020)
Lid interlocking system prevents accidental opening during operation; Natural rotor airflow minimizes heat buildup to protect samples; Brushless motor runs quietly (<60dB)
120 V, 50/60 Hz (US)
Keypad
14,000 to 14,999 rpm
No
Eppendorf MiniSpin / MiniSpin Plus
F-45-12-11
Videos
SDS
Documents
Documents
Accessories