Eppendorf™ PCR Tube Work Tray and Frame
Ideal for sample prep and transport
Supplier: Eppendorf™ 951010049
Description
- Polycarbonate work tray, plus frame (Mfr. No. 951010049), makes a benchtop rack that holds 96 × 0.2mL tubes
- Load tubes into rack and transfer the tray portion directly onto thermal cycler block
- Autoclavable
Specifications
Frame | |
Autoclavable | |
Polycarbonate |