Eppendorf™ PCR Tube Work Tray and Frame
Ideal for sample prep and transport
$154.50 - $154.50
Catalog Number | Mfr. No. | Product Type | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Product Type | Price | Quantity | |||||
E0030124308
|
Eppendorf™
951010049 |
Frame |
Case of 5 Each for $154.50
|
|
|||||
Description
- Polycarbonate work tray, plus frame (Mfr. No. 951010049), makes a benchtop rack that holds 96 × 0.2mL tubes
- Load tubes into rack and transfer the tray portion directly onto thermal cycler block
- Autoclavable
Specifications
Autoclavable | |
Polycarbonate |