Eppendorf™ Reference™ 2 Variable Volume Pipets- Trade-In Offer
Available in trade-in offer at reduced price
Supplier:  Eppendorf™ 2231302074
 View more versions of this product
Catalog No. 05-412-536
Explore available promotions
Includes: ep.T.I.P.S. Box
Description
Description
Exchange any brand pipet with brand new Reference 2 pipet (100μL to 2.5mL) at low costs
  • One-button operation pipet offers fast and ergonomic handling with reduced operating effort and active aerosol reduction
  • Reduced repetitive strain injuries by not having to move your thumb sideways
  • Spring loaded tip cone improves user reproducibility and ergonomics by lowering tip attachment and ejection force
  • Secondary user adjustment supports easy adjustment of pipet for accurate pipetting without need for full calibration
  • Four digit volume display with magnifying window for easier volume identification, volume lock feature prevents accidental changes in volume
  • Clear haptic feedback is felt after blowout step which avoids accidental tip ejections
  • Embedded RFID chip contains all relevant pipet data and TrackIT software for simple identification and documentation
  • Reference 2 multichannel pipets offers high throughput option including spring-loaded nose cones which can be switched on/off optionally
  • High flexibility with ability to remove single cones
  • Ensures continuous identical pipet alignment during work process
  • Serial number is printed on multiple components of pipet
  • Volume labeled pipet for quick identification of volume size/tip size
  • Round upper part of pipet makes it possible to work in every position
  • Sturdy upper handle, movable parts and external edges made from stainless-steel make pipet robust
  • Quick connect lower part enables easy cleaning and maintenance of pipet
  • Fully autoclavable pipet with smooth surface and few rough edges
Specifications
Specifications
Autoclavable
Reference™ 2
0.25 to 2.5 mL
Red
Pipette