Learn More
S'il vous plaît vous connecter à votre compte en ligne pour afficher votre prix réduits
Eppendorf™ Research™ plus Mechanical Multi-Channel Pipette
Adapt for specific liquids or other external conditions with these ultra-light fully autoclavable pipettes. Research™ plus Mechanical Multi-Channel Pipettes feature a spring loaded tip cone, a secondary adjustment option and an improved volume display.
$1852.95 - $3270.11
Spécifications
Autoclavable | Autoclavable |
---|---|
Type | Mechanical Pipette |
Display | 4 Digits |
Finger Style | Ergonomic |
Numéro de catalogue | Numéro du manufacturier. | Volume (Metric) | Number of Channels | Color | Includes | Prix | Quantité | ||
---|---|---|---|---|---|---|---|---|---|
Numéro de catalogue | Numéro du manufacturier. | Volume (Metric) | Number of Channels | Color | Includes | Prix | Quantité | ||
13-690-047
|
Eppendorf™
3125000010 |
0.5 to 10 μL | 8 | Medium Gray | ep.T.I.P.S.™ Box 2.0 with 96 Pipette Tips |
chaque for $1,955.69
|
|
||
13-690-048
|
Eppendorf™
3125000036 |
10 to 100 μL | 8 | Yellow | epT.I.P.S.™Â Box 2.0 with 96 Pipette Tips |
chaque for $1,852.95
|
|
||
13-690-049
|
Eppendorf™
3125000052 |
30 to 300 μL | 8 | Orange | epT.I.P.S.™Â Box 2.0 with 96 Pipette Tips |
chaque for $1,852.95
|
|
||
13-690-050
|
Eppendorf™
3125000028 |
0.5 to 10 μL | 12 | Medium Gray | epT.I.P.S.™Â Box 2.0 with 96 Pipette Tips |
chaque for $2,282.47
|
|
||
13-690-051
|
Eppendorf™
3125000044 |
10 to 100 μL | 12 | Yellow | epT.I.P.S.™Â Box 2.0 with 96 Pipette Tips |
chaque for $2,148.71
|
|
||
13-690-052
|
Eppendorf™
3125000060 |
30 to 300 μL | 12 | Orange | epT.I.P.S.™Â Box 2.0 with 96 Pipette Tips |
chaque for $2,148.71
|
|
||
05414270
|
Eppendorf™
3125000214* |
120 to 1200 μL | 8 | Dark Green | epT.I.P.S.™Â Box 2.0 with 96 Pipette Tips |
chaque for $2,822.26
|
|
||
05414271
|
Eppendorf™
3125000222* |
120 to 1200 μL | 12 | Dark Green | epT.I.P.S.™Â Box 2.0 with 96 Pipette Tips |
chaque for $3,270.11
|
|
||
05413602
|
Eppendorf™
3125000109 |
5 to 100 μL | 24 | Light Yellow | epT.I.P.S.™Â Box 2.0 with 384 Pipette Tips |
chaque for $2,574.29
|
|
||
05413600
|
Eppendorf™
3125000087 |
1 to 20 μL | 24 | Light Pink | epT.I.P.S.™Â Box 2.0 with 384 Pipette Tips |
chaque for $2,574.29
|
|
||
05413601
|
Eppendorf™
3125000095 |
5 to 100 μL | 16 | Light Yellow | epT.I.P.S.™Â Box 2.0 with 384 Pipette Tips |
chaque for $2,344.36
|
|
||
05413599
|
Eppendorf™
3125000079 |
1 to 20 μL | 16 | Light Pink | epT.I.P.S.™Â Box 2.0 with 384 Pipette Tips |
chaque for $2,344.36
|
|
||
Description
- Volume adjustment feature allows desired volume to be reached in few quick turns
- Control button and ejector's low operating force are positioned for ergonomics
- 4-digit, magnifying-shape volume display
- Calibration display and adjustment opening for specific liquids and volumes
- Uses renowned PhysioCare Concept™ thus reduces strain on hand and arm during pipetting, maintains balance, minimal user exertion
- Quick lock makes lower part particularly easy to remove and service (not available for 5 and 10 mL pipettes)
- Spring loaded tip cone promotes tight fit to tip (not available for 5 and 10 mL pipettes)
- Has ability to be autoclaved without disassembling
Applications
- Pipetting liquids
- Mixing liquids
- Filling of plates, gels, and reaction vessels
- Phase extraction and removal of supernatants
Spécifications
Autoclavable | |
Mechanical Pipette | |
4 Digits | |
Ergonomic |