Eppendorf™ epT.I.P.S.™ Sets - Pipette Tips
Eppendorf epT.I.P.S.™ Sets, in 'Eppendorf Quality' grade, combine the convenience of a reusable Box 2.0 with high-quality reloadable pipette tips, offering a sustainable, efficient solution for precise and organized pipetting in laboratories.
Supplier:  Eppendorf™ 0030076338
 View more versions of this product
Catalog No. 05414197
Explore available promotions
Description
Description
  • Contamination-free transfer of trays to stable working box
  • System optimized for use with multichannel pipettes
  • Color-coded trays for simple identification of tips and pipette
  • Tips can be attached to pipette from refill trays
  • The Reloads and Boxes can be autoclaved at 121 °C for subsequent use
  • epT.I.P.S. Box 2.0 and epT.I.P.S. Set available in Eppendorf Quality
  • ACT™-labeled by non-profit My Green Lab™ to help you make an informed product choice when it comes to sustainability
Specifications
Specifications
20 to 300 μL
Non-sterile
Autoclavable
Eppendorf Quality (Non-sterile)
Box
480 Pk.
Orange
Air-cushion System
epT.I.P.S.
Non-filtered
Colorless
epTIPS
Eppendorf Pipettes, Non-Eppendorf Pipettes
480 Tips (5 Trays x 96 Tips), 1 Reusable Box
Pipette Tips
Single-use
Yes
Polypropylene