missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Erbium(III) 2,4-pentanedionate, REacton™, 99.9% (REO), Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 01321404

 View more versions of this product

Catalog No. AA1321404


Explore available promotions

Description

Description

Erbium(III) 2,4-pentanedionate is commonly used in various catalysts and catalytic reagents for organic synthesis. It is also used for glass coloring, as an amplifier in fiber optics, and in lasers for medical and dental purposes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Erbium(III) 2, 4-pentanedionate
Powder
99.9% (REO)
MFCD00013490
Insoluble in water.
[Er+3].CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O
464.59
464.59
Hygroscopic
14553-08-3
2 g
C15H21ErO6
erbium 2,4-pentanedionate, erbium iii acetylacetonate hydrate, erbium iii 2,4-pentanedionate, reacton, 3z-4-bis 2z-4-oxopent-2-en-2-yl oxy erbio oxy pent-3-en-2-one
FGQSJRDKBCVFHH-UHFFFAOYSA-N
erbium(3+) tris(2,4-dioxopentan-3-ide)
102601666
Odorless

Chemical Identifiers

14553-08-3
464.59
FGQSJRDKBCVFHH-UHFFFAOYSA-N
102601666
C15H21ErO6
MFCD00013490
erbium 2,4-pentanedionate, erbium iii acetylacetonate hydrate, erbium iii 2,4-pentanedionate, reacton, 3z-4-bis 2z-4-oxopent-2-en-2-yl oxy erbio oxy pent-3-en-2-one
[Er+3].CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O
Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only