missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Erbium(III) 2,4-pentanedionate, REacton™, 99.9% (REO), Thermo Scientific Chemicals
$186.63 - $785.70
Chemical Identifiers
CAS | 14553-08-3 |
---|---|
Molecular Formula | C15H21ErO6 |
Molecular Weight (g/mol) | 464.59 |
MDL Number | MFCD00013490 |
InChI Key | FGQSJRDKBCVFHH-UHFFFAOYSA-N |
Synonym | erbium 2,4-pentanedionate, erbium iii acetylacetonate hydrate, erbium iii 2,4-pentanedionate, reacton, 3z-4-bis 2z-4-oxopent-2-en-2-yl oxy erbio oxy pent-3-en-2-one |
PubChem CID | 102601666 |
SMILES | [Er+3].CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O |
Description
Erbium(III) 2,4-pentanedionate is commonly used in various catalysts and catalytic reagents for organic synthesis. It is also used for glass coloring, as an amplifier in fiber optics, and in lasers for medical and dental purposes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
14553-08-3 | |
464.59 | |
FGQSJRDKBCVFHH-UHFFFAOYSA-N | |
102601666 |
C15H21ErO6 | |
MFCD00013490 | |
erbium 2,4-pentanedionate, erbium iii acetylacetonate hydrate, erbium iii 2,4-pentanedionate, reacton, 3z-4-bis 2z-4-oxopent-2-en-2-yl oxy erbio oxy pent-3-en-2-one | |
[Er+3].CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O |
Specifications
14553-08-3 | |
2 g | |
C15H21ErO6 | |
erbium 2,4-pentanedionate, erbium iii acetylacetonate hydrate, erbium iii 2,4-pentanedionate, reacton, 3z-4-bis 2z-4-oxopent-2-en-2-yl oxy erbio oxy pent-3-en-2-one | |
FGQSJRDKBCVFHH-UHFFFAOYSA-N | |
erbium(3+) tris(2,4-dioxopentan-3-ide) | |
102601666 | |
Odorless | |
Erbium(III) 2, 4-pentanedionate |
Powder | |
99.9% (REO) | |
MFCD00013490 | |
Insoluble in water. | |
[Er+3].CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O | |
464.59 | |
464.59 | |
Hygroscopic |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only