missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Erbium(III) isopropoxide, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 03661303

 View more versions of this product

Catalog No. AA3661303


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Erbium(III) isopropoxide
325°C (decomposition)
Alcohol-like
Er[OCH(CH3)2]3
1 g
Air and moisture sensitive
VOCNVTAXVORJBI-UHFFFAOYSA-N
erbium(3+);propan-2-olate
2734042
Powder
14814-07-4
58°C (138°F)
C9H21ErO3
MFCD00144253
UN2925
erbium iii isopropoxide, triisopropoxyerbium, erbium-isopropoxide, acmc-20al30, erbium iii i-propoxide, erbium 3+ ; propan-2-olate, erbium 3+ ion tris propan-2-olate, erbium 3+ tris propan-2-olate
CC(C)[O-].CC(C)[O-].CC(C)[O-].[Er+3]
344.523
344.52

Chemical Identifiers

14814-07-4
344.523
VOCNVTAXVORJBI-UHFFFAOYSA-N
2734042
CC(C)[O-].CC(C)[O-].CC(C)[O-].[Er+3]
C9H21ErO3
MFCD00144253
erbium iii isopropoxide, triisopropoxyerbium, erbium-isopropoxide, acmc-20al30, erbium iii i-propoxide, erbium 3+ ; propan-2-olate, erbium 3+ ion tris propan-2-olate, erbium 3+ tris propan-2-olate
erbium(3+);propan-2-olate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H228
Causes severe skin burns and eye damage.
Flammable solid.

P210-P240-P241-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c

H228-H314

DOTInformation : Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: FLAMMABLE SOLIDS, CORROSIVE, ORGANIC, N.O.S.

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only