Learn More
Erbium(III) isopropoxide, Thermo Scientific Chemicals
$121.85 - $446.60
Chemical Identifiers
CAS | 14814-07-4 |
---|---|
Molecular Formula | C9H21ErO3 |
Molecular Weight (g/mol) | 344.523 |
MDL Number | MFCD00144253 |
InChI Key | VOCNVTAXVORJBI-UHFFFAOYSA-N |
Synonym | erbium iii isopropoxide, triisopropoxyerbium, erbium-isopropoxide, acmc-20al30, erbium iii i-propoxide, erbium 3+ ; propan-2-olate, erbium 3+ ion tris propan-2-olate, erbium 3+ tris propan-2-olate |
PubChem CID | 2734042 |
IUPAC Name | erbium(3+);propan-2-olate |
SMILES | CC(C)[O-].CC(C)[O-].CC(C)[O-].[Er+3] |
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
14814-07-4 | |
344.523 | |
VOCNVTAXVORJBI-UHFFFAOYSA-N | |
2734042 | |
CC(C)[O-].CC(C)[O-].CC(C)[O-].[Er+3] |
C9H21ErO3 | |
MFCD00144253 | |
erbium iii isopropoxide, triisopropoxyerbium, erbium-isopropoxide, acmc-20al30, erbium iii i-propoxide, erbium 3+ ; propan-2-olate, erbium 3+ ion tris propan-2-olate, erbium 3+ tris propan-2-olate | |
erbium(3+);propan-2-olate |
Specifications
14814-07-4 | |
58°C (138°F) | |
C9H21ErO3 | |
MFCD00144253 | |
UN2925 | |
erbium iii isopropoxide, triisopropoxyerbium, erbium-isopropoxide, acmc-20al30, erbium iii i-propoxide, erbium 3+ ; propan-2-olate, erbium 3+ ion tris propan-2-olate, erbium 3+ tris propan-2-olate | |
CC(C)[O-].CC(C)[O-].CC(C)[O-].[Er+3] | |
344.523 | |
344.52 | |
Erbium(III) isopropoxide |
325°C (decomposition) | |
Alcohol-like | |
Er[OCH(CH3)2]3 | |
1 g | |
Air and moisture sensitive | |
VOCNVTAXVORJBI-UHFFFAOYSA-N | |
erbium(3+);propan-2-olate | |
2734042 | |
Powder |
Safety and Handling
GHS H Statement
H314-H228
Causes severe skin burns and eye damage.
Flammable solid.
P210-P240-P241-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
H228-H314
DOTInformation : Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: FLAMMABLE SOLIDS, CORROSIVE, ORGANIC, N.O.S.
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only