missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Impacto™ Ultra Work Sport Anti-Fatigue Insoles
Impacto™ ERINWRK Ultra Work Sport Insoles help protect against back and knee pain and impact-related injuries such as plantar fasciitis, Achilles tendonitis, runner's knee and shin splints.
Supplier: Impacto™ ERINWRKD
Description
Impacto™ Ultra Work-Sport Anti-Fatigue Insoles
- Sorbothane™ inlays absorb shock and help relieve pain in feet, knees and lower back
- Designed to provide cushioned support and shock absorbing comfort
- Cupped heel for rear foot stability
- Breathable and antifungal
- For use with closed shoes and boots
Specifications
Blue | |
27.94 cm | |
Men: 8 to 9; Women: 10.5 to 11.5 | |
Ergotech Ultra Work Sport Insoles |
11 in. | |
Sorbothane | |
Unisex |