missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing
Ethyl 2,4-dioxo-4-(2-thienyl)butanoate, 90%, Thermo Scientific™
Chemical Identifiers
CAS |
36983-36-5 |
Molecular Formula |
C10H10O4S |
Molecular Weight (g/mol) |
226.246 |
MDL Number |
MFCD00126316 |
InChI Key |
GCCFYXYKJZUHMI-UHFFFAOYSA-N |
Synonym |
ethyl 2,4-dioxo-4-thiophen-2-yl butanoate, ethyl 2,4-dioxo-4-2-thienyl butanoate, 2,4-dioxo-4-thiophen-2-yl-butyric acid ethyl ester, 2-thiophenebutanoicacid, a,g-dioxo-, ethyl ester, 2,4-dioxo-4-2-thienyl butanoic acid ethyl ester, 2,4-dioxo-4-thiophen-2-ylbutyric acid ethyl ester |
PubChem CID |
2799459 |
IUPAC Name |
ethyl 2,4-dioxo-4-thiophen-2-ylbutanoate |
SMILES |
CCOC(=O)C(=O)CC(=O)C1=CC=CS1 |
View More Specs
Products
1
Catalog Number |
Mfr. No. |
Quantity |
Packaging |
Price |
Quantity |
|
|
|
|
SPB04027EA
SDS
View Documents
|
Thermo Scientific Chemicals
SPB04027EA
|
10 g |
Amber Glass Bottle |
N/A
|
|
Add to cart
|
|
|
|
Description
Specifications
Chemical Identifiers
|
36983-36-5 |
|
226.246 |
|
GCCFYXYKJZUHMI-UHFFFAOYSA-N |
|
2799459 |
|
CCOC(=O)C(=O)CC(=O)C1=CC=CS1 |
|
C10H10O4S |
|
MFCD00126316 |
|
ethyl 2,4-dioxo-4-thiophen-2-yl butanoate, ethyl 2,4-dioxo-4-2-thienyl butanoate, 2,4-dioxo-4-thiophen-2-yl-butyric acid ethyl ester, 2-thiophenebutanoicacid, a,g-dioxo-, ethyl ester, 2,4-dioxo-4-2-thienyl butanoic acid ethyl ester, 2,4-dioxo-4-thiophen-2-ylbutyric acid ethyl ester |
|
ethyl 2,4-dioxo-4-thiophen-2-ylbutanoate |
Specifications
|
36983-36-5 |
|
90% |
|
C10H10O4S |
|
10 g |
|
GCCFYXYKJZUHMI-UHFFFAOYSA-N |
|
ethyl 2,4-dioxo-4-thiophen-2-ylbutanoate |
|
2799459 |
|
90% |
|
90.0 |
|
Amber Glass Bottle |
|
MFCD00126316 |
|
ethyl 2,4-dioxo-4-thiophen-2-yl butanoate, ethyl 2,4-dioxo-4-2-thienyl butanoate, 2,4-dioxo-4-thiophen-2-yl-butyric acid ethyl ester, 2-thiophenebutanoicacid, a,g-dioxo-, ethyl ester, 2,4-dioxo-4-2-thienyl butanoic acid ethyl ester, 2,4-dioxo-4-thiophen-2-ylbutyric acid ethyl ester |
|
CCOC(=O)C(=O)CC(=O)C1=CC=CS1 |
|
226.246 |
|
226.25 |
|
Ethyl 2, 4-dioxo-4-(2-thienyl)butanoate |
Videos
SDS
missing translation for 'documents'