missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethyl 2-amidinoacetate hydrochloride, 95%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals H6428814

 View more versions of this product

Catalog No. AAH6428814


Explore available promotions

Description

Description

It is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a pharmaceutical intermediate.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications

Specifications

Ethyl 2-amidinoacetate hydrochloride
105°C to 108°C
MFCD06797615
3694030
VOHFLYOSVGWQOS-UHFFFAOYSA-N
ethyl 3-amino-3-iminopropanoate;hydrochloride
15555354
95%
57508-48-2
C5H11ClN2O2
25 g
ethyl 3-amino-3-iminopropanoate hydrochloride, ethyl 2-amidinoacetate hydrochloride, 3-amino-3-iminopropanoic acid ethyl ester hydrochloride, ethyl 2-carbamimidoylacetate hydrochloride, 3-amino-3-imino-propanoic acid, ethyl ester hcl, carbamimidoyl-acetic acid ethyl ester hydrochloride, ethyl 2-amidinoacetate hcl, ethyl 3-amino-3-iminopropanate hydrochloride, ethyl 3-amino-3-iminopropanoate hcl, 3-amino-3-iminopropionic acid ethyl ester hydrochloride
CCOC(=O)CC(=N)N.Cl
166.605
166.6
Solid

Chemical Identifiers

57508-48-2
166.605
VOHFLYOSVGWQOS-UHFFFAOYSA-N
15555354
CCOC(=O)CC(=N)N.Cl
C5H11ClN2O2
MFCD06797615
ethyl 3-amino-3-iminopropanoate hydrochloride, ethyl 2-amidinoacetate hydrochloride, 3-amino-3-iminopropanoic acid ethyl ester hydrochloride, ethyl 2-carbamimidoylacetate hydrochloride, 3-amino-3-imino-propanoic acid, ethyl ester hcl, carbamimidoyl-acetic acid ethyl ester hydrochloride, ethyl 2-amidinoacetate hcl, ethyl 3-amino-3-iminopropanate hydrochloride, ethyl 3-amino-3-iminopropanoate hcl, 3-amino-3-iminopropionic acid ethyl ester hydrochloride
ethyl 3-amino-3-iminopropanoate;hydrochloride
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only