missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethyl 2-amidinoacetate hydrochloride, 95%, Thermo Scientific Chemicals

$184.96 - $888.74

Chemical Identifiers

CAS 57508-48-2
Molecular Formula C5H11ClN2O2
Molecular Weight (g/mol) 166.605
MDL Number MFCD06797615
InChI Key VOHFLYOSVGWQOS-UHFFFAOYSA-N
Synonym ethyl 3-amino-3-iminopropanoate hydrochloride, ethyl 2-amidinoacetate hydrochloride, 3-amino-3-iminopropanoic acid ethyl ester hydrochloride, ethyl 2-carbamimidoylacetate hydrochloride, 3-amino-3-imino-propanoic acid, ethyl ester hcl, carbamimidoyl-acetic acid ethyl ester hydrochloride, ethyl 2-amidinoacetate hcl, ethyl 3-amino-3-iminopropanate hydrochloride, ethyl 3-amino-3-iminopropanoate hcl, 3-amino-3-iminopropionic acid ethyl ester hydrochloride
PubChem CID 15555354
IUPAC Name ethyl 3-amino-3-iminopropanoate;hydrochloride
SMILES CCOC(=O)CC(=N)N.Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6428806
View Documents
Thermo Scientific Chemicals
H6428806
5 g
Each for $184.96
 
AAH6428814
View Documents
Thermo Scientific Chemicals
H6428814
25 g
Each for $888.74
 
Description

Description

It is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a pharmaceutical intermediate.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications

Chemical Identifiers

57508-48-2
166.605
VOHFLYOSVGWQOS-UHFFFAOYSA-N
15555354
CCOC(=O)CC(=N)N.Cl
C5H11ClN2O2
MFCD06797615
ethyl 3-amino-3-iminopropanoate hydrochloride, ethyl 2-amidinoacetate hydrochloride, 3-amino-3-iminopropanoic acid ethyl ester hydrochloride, ethyl 2-carbamimidoylacetate hydrochloride, 3-amino-3-imino-propanoic acid, ethyl ester hcl, carbamimidoyl-acetic acid ethyl ester hydrochloride, ethyl 2-amidinoacetate hcl, ethyl 3-amino-3-iminopropanate hydrochloride, ethyl 3-amino-3-iminopropanoate hcl, 3-amino-3-iminopropionic acid ethyl ester hydrochloride
ethyl 3-amino-3-iminopropanoate;hydrochloride

Specifications

57508-48-2
C5H11ClN2O2
5 g
ethyl 3-amino-3-iminopropanoate hydrochloride, ethyl 2-amidinoacetate hydrochloride, 3-amino-3-iminopropanoic acid ethyl ester hydrochloride, ethyl 2-carbamimidoylacetate hydrochloride, 3-amino-3-imino-propanoic acid, ethyl ester hcl, carbamimidoyl-acetic acid ethyl ester hydrochloride, ethyl 2-amidinoacetate hcl, ethyl 3-amino-3-iminopropanate hydrochloride, ethyl 3-amino-3-iminopropanoate hcl, 3-amino-3-iminopropionic acid ethyl ester hydrochloride
CCOC(=O)CC(=N)N.Cl
166.605
166.6
Solid
105°C to 108°C
MFCD06797615
3694030
VOHFLYOSVGWQOS-UHFFFAOYSA-N
ethyl 3-amino-3-iminopropanoate;hydrochloride
15555354
95%
Ethyl 2-amidinoacetate hydrochloride
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only