missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethyl 2-bromoisovalerate, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals B2252514

 View more versions of this product

Catalog No. AAB2252514


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Ethyl 2-bromoisovalerate
1.276
65°C (149°F)
C7H13BrO2
MFCD00026855
UN3265
ethyl 2-bromoisovalerate, ethyl 2-bromo isovalerate, ethyl 2-bromo-3-methylbutyrate, ethyl .alpha.-bromoisovalerate, ethyl .alpha.-bromoisovalerate, 2-bromoisovaleric acid ethyl ester, ethyl 2-bromo-3-methyl-butanoate, .alpha.-bromoisovaleric acid ethyl ester, ethyl, a-bromoisovalerate, a-bromoisovalerate
CCOC(=O)C(C(C)C)Br
209.083
209.09
609-12-1
185°C to 186°C
Pungent
1.449
25 g
1099039
WNFUWONOILPKNX-UHFFFAOYSA-N
ethyl 2-bromo-3-methylbutanoate
79068
97%

Chemical Identifiers

609-12-1
209.083
WNFUWONOILPKNX-UHFFFAOYSA-N
79068
CCOC(=O)C(C(C)C)Br
C7H13BrO2
MFCD00026855
ethyl 2-bromoisovalerate, ethyl 2-bromo isovalerate, ethyl 2-bromo-3-methylbutyrate, ethyl .alpha.-bromoisovalerate, ethyl .alpha.-bromoisovalerate, 2-bromoisovaleric acid ethyl ester, ethyl 2-bromo-3-methyl-butanoate, .alpha.-bromoisovaleric acid ethyl ester, ethyl, a-bromoisovalerate, a-bromoisovalerate
ethyl 2-bromo-3-methylbutanoate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.

P210-P280-P370+P378q-P501c

H227

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.

EINECSNumber : 210-178-3

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only