missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethyl 2-bromoisovalerate, 97%, Thermo Scientific Chemicals

$140.99 - $140.99

Chemical Identifiers

CAS 609-12-1
Molecular Formula C7H13BrO2
Molecular Weight (g/mol) 209.083
MDL Number MFCD00026855
InChI Key WNFUWONOILPKNX-UHFFFAOYSA-N
Synonym ethyl 2-bromoisovalerate, ethyl 2-bromo isovalerate, ethyl 2-bromo-3-methylbutyrate, ethyl .alpha.-bromoisovalerate, ethyl .alpha.-bromoisovalerate, 2-bromoisovaleric acid ethyl ester, ethyl 2-bromo-3-methyl-butanoate, .alpha.-bromoisovaleric acid ethyl ester, ethyl, a-bromoisovalerate, a-bromoisovalerate
PubChem CID 79068
IUPAC Name ethyl 2-bromo-3-methylbutanoate
SMILES CCOC(=O)C(C(C)C)Br
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2252514
View Documents
Thermo Scientific Chemicals
B2252514
25 g
Each for $140.99
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

609-12-1
209.083
WNFUWONOILPKNX-UHFFFAOYSA-N
79068
CCOC(=O)C(C(C)C)Br
C7H13BrO2
MFCD00026855
ethyl 2-bromoisovalerate, ethyl 2-bromo isovalerate, ethyl 2-bromo-3-methylbutyrate, ethyl .alpha.-bromoisovalerate, ethyl .alpha.-bromoisovalerate, 2-bromoisovaleric acid ethyl ester, ethyl 2-bromo-3-methyl-butanoate, .alpha.-bromoisovaleric acid ethyl ester, ethyl, a-bromoisovalerate, a-bromoisovalerate
ethyl 2-bromo-3-methylbutanoate

Specifications

609-12-1
185°C to 186°C
Pungent
1.449
25 g
1099039
WNFUWONOILPKNX-UHFFFAOYSA-N
ethyl 2-bromo-3-methylbutanoate
79068
97%
1.276
65°C (149°F)
C7H13BrO2
MFCD00026855
UN3265
ethyl 2-bromoisovalerate, ethyl 2-bromo isovalerate, ethyl 2-bromo-3-methylbutyrate, ethyl .alpha.-bromoisovalerate, ethyl .alpha.-bromoisovalerate, 2-bromoisovaleric acid ethyl ester, ethyl 2-bromo-3-methyl-butanoate, .alpha.-bromoisovaleric acid ethyl ester, ethyl, a-bromoisovalerate, a-bromoisovalerate
CCOC(=O)C(C(C)C)Br
209.083
209.09
Ethyl 2-bromoisovalerate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.

P210-P280-P370+P378q-P501c

H227

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.

EINECSNumber : 210-178-3

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only