Learn More
Ethyl 3,4-dihydroxybenzoate, 98%, Thermo Scientific Chemicals
$87.07 - $1373.36
Chemical Identifiers
CAS | 3943-89-3 |
---|---|
Molecular Formula | C9H10O4 |
Molecular Weight (g/mol) | 182.18 |
MDL Number | MFCD00002199 |
InChI Key | KBPUBCVJHFXPOC-UHFFFAOYSA-N |
Synonym | ethyl protocatechuate, 3,4-dihydroxybenzoic acid ethyl ester, ethyl-3,4-dihydroxybenzoate, unii-4ygj96wtbg, benzoic acid, 3,4-dihydroxy-, ethyl ester, edhb, protocatechuic acid ethyl ester, 4ygj96wtbg, 3,4-dihydroxy-benzoic acid ethyl ester |
PubChem CID | 77547 |
IUPAC Name | ethyl 3,4-dihydroxybenzoate |
SMILES | CCOC(=O)C1=CC=C(O)C(O)=C1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1100109
|
Thermo Scientific Chemicals
A1100109 |
10 g |
Each for $87.07
|
|
|||||
AAA1100118
|
Thermo Scientific Chemicals
A1100118 |
50 g |
Each for $338.99
|
|
|||||
AAA1100130
|
Thermo Scientific Chemicals
A1100130 |
250 g |
Each for $1,373.36
|
|
|||||
Description
The compound is a prolyl 4-hydroxylase inhibitor and can be used to protect the myocardium.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsThe compound is a prolyl 4-hydroxylase inhibitor and can be used to protect the myocardium.
Solubility
Insoluble in water. Soluble in ethanol.
Notes
Keep container tightly closed in a cool, dry and well-ventilated place. Store away from oxidizing agents, Strong bases.
Chemical Identifiers
3943-89-3 | |
182.18 | |
KBPUBCVJHFXPOC-UHFFFAOYSA-N | |
77547 | |
CCOC(=O)C1=CC=C(O)C(O)=C1 |
C9H10O4 | |
MFCD00002199 | |
ethyl protocatechuate, 3,4-dihydroxybenzoic acid ethyl ester, ethyl-3,4-dihydroxybenzoate, unii-4ygj96wtbg, benzoic acid, 3,4-dihydroxy-, ethyl ester, edhb, protocatechuic acid ethyl ester, 4ygj96wtbg, 3,4-dihydroxy-benzoic acid ethyl ester | |
ethyl 3,4-dihydroxybenzoate |
Specifications
3943-89-3 | |
C9H10O4 | |
10 g | |
ethyl protocatechuate, 3,4-dihydroxybenzoic acid ethyl ester, ethyl-3,4-dihydroxybenzoate, unii-4ygj96wtbg, benzoic acid, 3,4-dihydroxy-, ethyl ester, edhb, protocatechuic acid ethyl ester, 4ygj96wtbg, 3,4-dihydroxy-benzoic acid ethyl ester | |
KBPUBCVJHFXPOC-UHFFFAOYSA-N | |
ethyl 3,4-dihydroxybenzoate | |
77547 | |
98% |
132°C to 135°C | |
MFCD00002199 | |
2097435 | |
Insoluble in water. Soluble in ethanol. | |
CCOC(=O)C1=CC=C(O)C(O)=C1 | |
182.18 | |
182.18 | |
Ethyl 3,4-dihydroxybenzoate |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 223-529-0
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only