missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethyl 3-bromopropionate, 99%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1240222

 View more versions of this product

Catalog No. AAA1240222

Explore available promotions

Description

Description

Ethyl 3-bromopropionate was used in the preparation of (3?,4?-dimethoxyphenyl)-5-oxopentanoic acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl 3-bromopropionate was used in the preparation of (3′,4′-dimethoxyphenyl)-5-oxopentanoic acid.

Solubility
Soluble in water, alcohol, chloroform, benzene.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, Bases, Reducing agents
Specifications

Specifications

Ethyl 3-bromopropionate
1.416
79°C (174°F)
1.452
100 g
Light sensitive
Soluble in water,alcohol,chloroform,benzene.
CCOC(=O)CCBr
181.03
181.03
539-74-2
67°C to 71°C (10 mmHg)
C5H9BrO2
MFCD00000251
1700933
ethyl 3-bromopropionate, propanoic acid, 3-bromo-, ethyl ester, 3-bromopropionic acid ethyl ester, ethyl beta-bromopropionate, unii-9b28g9s1jv, ccris 9051, propionic acid, 3-bromo-, ethyl ester, ethyl .beta.-bromopropionate, 3-bromo-propionic acid ethyl ester, ethyl-3-bromopropionate
FQTIYMRSUOADDK-UHFFFAOYSA-N
ethyl 3-bromopropanoate
68320
99%

Chemical Identifiers

539-74-2
181.03
FQTIYMRSUOADDK-UHFFFAOYSA-N
68320
CCOC(=O)CCBr
C5H9BrO2
MFCD00000251
ethyl 3-bromopropionate, propanoic acid, 3-bromo-, ethyl ester, 3-bromopropionic acid ethyl ester, ethyl beta-bromopropionate, unii-9b28g9s1jv, ccris 9051, propionic acid, 3-bromo-, ethyl ester, ethyl .beta.-bromopropionate, 3-bromo-propionic acid ethyl ester, ethyl-3-bromopropionate
ethyl 3-bromopropanoate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.

P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c

H227-H315-H319-H335

EINECSNumber : 208-724-0

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only