Learn More
Ethyl 3-bromopropionate, 99%, Thermo Scientific Chemicals
$121.07 - $1,577.82
Chemical Identifiers
CAS | 539-74-2 |
---|---|
Molecular Formula | C5H9BrO2 |
Molecular Weight (g/mol) | 181.03 |
MDL Number | MFCD00000251 |
InChI Key | FQTIYMRSUOADDK-UHFFFAOYSA-N |
Synonym | ethyl 3-bromopropionate, propanoic acid, 3-bromo-, ethyl ester, 3-bromopropionic acid ethyl ester, ethyl beta-bromopropionate, unii-9b28g9s1jv, ccris 9051, propionic acid, 3-bromo-, ethyl ester, ethyl .beta.-bromopropionate, 3-bromo-propionic acid ethyl ester, ethyl-3-bromopropionate |
PubChem CID | 68320 |
IUPAC Name | ethyl 3-bromopropanoate |
SMILES | CCOC(=O)CCBr |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
AAA1240214
|
Thermo Scientific Chemicals
A1240214 |
25 g |
N/A
|
|
||||||
AAA1240222
|
Thermo Scientific Chemicals
A1240222 |
100 g |
N/A
|
|
||||||
AAA1240236
|
Thermo Scientific Chemicals
A1240236 |
500 g |
N/A
|
|
||||||
Description
Ethyl 3-bromopropionate was used in the preparation of (3?,4?-dimethoxyphenyl)-5-oxopentanoic acid.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsEthyl 3-bromopropionate was used in the preparation of (3′,4′-dimethoxyphenyl)-5-oxopentanoic acid.
Solubility
Soluble in water, alcohol, chloroform, benzene.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, Bases, Reducing agents
Chemical Identifiers
539-74-2 | |
181.03 | |
FQTIYMRSUOADDK-UHFFFAOYSA-N | |
68320 | |
CCOC(=O)CCBr |
C5H9BrO2 | |
MFCD00000251 | |
ethyl 3-bromopropionate, propanoic acid, 3-bromo-, ethyl ester, 3-bromopropionic acid ethyl ester, ethyl beta-bromopropionate, unii-9b28g9s1jv, ccris 9051, propionic acid, 3-bromo-, ethyl ester, ethyl .beta.-bromopropionate, 3-bromo-propionic acid ethyl ester, ethyl-3-bromopropionate | |
ethyl 3-bromopropanoate |
Specifications
539-74-2 | |
67°C to 71°C (10 mmHg) | |
C5H9BrO2 | |
MFCD00000251 | |
1700933 | |
ethyl 3-bromopropionate, propanoic acid, 3-bromo-, ethyl ester, 3-bromopropionic acid ethyl ester, ethyl beta-bromopropionate, unii-9b28g9s1jv, ccris 9051, propionic acid, 3-bromo-, ethyl ester, ethyl .beta.-bromopropionate, 3-bromo-propionic acid ethyl ester, ethyl-3-bromopropionate | |
FQTIYMRSUOADDK-UHFFFAOYSA-N | |
ethyl 3-bromopropanoate | |
68320 | |
99% |
1.416 | |
79°C (174°F) | |
1.452 | |
25 g | |
Light sensitive | |
Soluble in water,alcohol,chloroform,benzene. | |
CCOC(=O)CCBr | |
181.03 | |
181.03 | |
Ethyl 3-bromopropionate |
Safety and Handling
GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
H227-H315-H319-H335
EINECSNumber : 208-724-0
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only