missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethyl 3-ethyl-5-methylisoxazole-4-carboxylate, 98%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals L0052606

 View more versions of this product

Catalog No. AAL0052606


Explore available promotions

Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

Ethyl 3-ethyl-5-methylisoxazole-4-carboxylate
1.07
103°C (217°F)
1.463
5 g
ethyl 3-ethyl-5-methylisoxazole-4-carboxylate, ethyl 3-ethyl-5-methyl-4-isoxazolecarboxylate, 3-ethyl-5-methyl-4-isoxazolecarboxylicacid ethylester, 3-ethyl-5-methyl-isoxazole-4-carboxylic acid ethyl ester, acmc-1b03d, 4-isoxazolecarboxylic acid, 3-ethyl-5-methyl-, ethyl ester
CCC1=NOC(=C1C(=O)OCC)C
183.207
183.21
53064-41-8
71°C to 72°C (0.5 mmHg)
C9H13NO3
MFCD00014094
1073370
JDWLDZUUTIPZHV-UHFFFAOYSA-N
ethyl 3-ethyl-5-methyl-1,2-oxazole-4-carboxylate
3745090
98%

Chemical Identifiers

53064-41-8
183.207
JDWLDZUUTIPZHV-UHFFFAOYSA-N
3745090
CCC1=NOC(=C1C(=O)OCC)C
C9H13NO3
MFCD00014094
ethyl 3-ethyl-5-methylisoxazole-4-carboxylate, ethyl 3-ethyl-5-methyl-4-isoxazolecarboxylate, 3-ethyl-5-methyl-4-isoxazolecarboxylicacid ethylester, 3-ethyl-5-methyl-isoxazole-4-carboxylic acid ethyl ester, acmc-1b03d, 4-isoxazolecarboxylic acid, 3-ethyl-5-methyl-, ethyl ester
ethyl 3-ethyl-5-methyl-1,2-oxazole-4-carboxylate
Safety and Handling

Safety and Handling

EINECSNumber : 000-000-0

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions