missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Ethyl 3-ethyl-5-methylisoxazole-4-carboxylate, 98%, Thermo Scientific™
$121.71 - $121.71
Chemical Identifiers
CAS | 53064-41-8 |
---|---|
Molecular Formula | C9H13NO3 |
Molecular Weight (g/mol) | 183.207 |
MDL Number | MFCD00014094 |
InChI Key | JDWLDZUUTIPZHV-UHFFFAOYSA-N |
Synonym | ethyl 3-ethyl-5-methylisoxazole-4-carboxylate, ethyl 3-ethyl-5-methyl-4-isoxazolecarboxylate, 3-ethyl-5-methyl-4-isoxazolecarboxylicacid ethylester, 3-ethyl-5-methyl-isoxazole-4-carboxylic acid ethyl ester, acmc-1b03d, 4-isoxazolecarboxylic acid, 3-ethyl-5-methyl-, ethyl ester |
PubChem CID | 3745090 |
IUPAC Name | ethyl 3-ethyl-5-methyl-1,2-oxazole-4-carboxylate |
SMILES | CCC1=NOC(=C1C(=O)OCC)C |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAL0052606
|
Thermo Scientific Chemicals
L0052606 |
5 g |
N/A
|
|
|||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
53064-41-8 | |
183.207 | |
JDWLDZUUTIPZHV-UHFFFAOYSA-N | |
3745090 | |
CCC1=NOC(=C1C(=O)OCC)C |
C9H13NO3 | |
MFCD00014094 | |
ethyl 3-ethyl-5-methylisoxazole-4-carboxylate, ethyl 3-ethyl-5-methyl-4-isoxazolecarboxylate, 3-ethyl-5-methyl-4-isoxazolecarboxylicacid ethylester, 3-ethyl-5-methyl-isoxazole-4-carboxylic acid ethyl ester, acmc-1b03d, 4-isoxazolecarboxylic acid, 3-ethyl-5-methyl-, ethyl ester | |
ethyl 3-ethyl-5-methyl-1,2-oxazole-4-carboxylate |
Specifications
53064-41-8 | |
71°C to 72°C (0.5 mmHg) | |
C9H13NO3 | |
MFCD00014094 | |
1073370 | |
JDWLDZUUTIPZHV-UHFFFAOYSA-N | |
ethyl 3-ethyl-5-methyl-1,2-oxazole-4-carboxylate | |
3745090 | |
98% |
1.07 | |
103°C (217°F) | |
1.463 | |
5 g | |
ethyl 3-ethyl-5-methylisoxazole-4-carboxylate, ethyl 3-ethyl-5-methyl-4-isoxazolecarboxylate, 3-ethyl-5-methyl-4-isoxazolecarboxylicacid ethylester, 3-ethyl-5-methyl-isoxazole-4-carboxylic acid ethyl ester, acmc-1b03d, 4-isoxazolecarboxylic acid, 3-ethyl-5-methyl-, ethyl ester | |
CCC1=NOC(=C1C(=O)OCC)C | |
183.207 | |
183.21 | |
Ethyl 3-ethyl-5-methylisoxazole-4-carboxylate |
Safety and Handling
EINECSNumber : 000-000-0
TSCA : No
Recommended Storage : Ambient temperatures