Learn More
Ethyl 3-isocyanatopropionate, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals L0961606
Description
Ethyl 3-isocyanatopropionate can be used to produce 3-[1-(2-dimethylamino-ethylcarbamoyl)-5,6-dimethyl-6H-pyrido[4,3-b]carbazol-9-yloxycarbonylamino]-propionic acid ethyl ester with 9-hydroxy-5,6-dimethyl-6H-pyrido[4,3-b]carbazole-1-carboxylic acid (2-dimethylamino-ethyl)-amide.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsEthyl 3-isocyanatopropionate can be used to produce 3-[1-(2-dimethylamino-ethylcarbamoyl)-5,6-dimethyl-6H-pyrido[4,3-b]carbazol-9-yloxycarbonylamino]-propionic acid ethyl ester with 9-hydroxy-5,6-dimethyl-6H-pyrido[4,3-b]carbazole-1-carboxylic acid (2-dimethylamino-ethyl)-amide.
Solubility
Not miscible with water.
Notes
Store at 4°C. Moisture Sensitive. Avoid moisture, heat and direct sunlight. Incompatible materials are amines, alcohols and heat. Store away from moisture and oxidizing agents.
Specifications
Ethyl 3-isocyanatopropionate | |
1.087 | |
92°C (197°F) | |
C6H9NO3 | |
MFCD00078461 | |
UN2206 | |
Moisture sensitive | |
Not miscible with water. | |
CCOC(=O)CCN=C=O | |
143.14 | |
143.14 |
5100-34-5 | |
74°C to 75°C (4 mmHg) | |
Irritating | |
1.429 | |
5 g | |
1705618 | |
ethyl 3-isocyanatopropionate, propanoic acid, 3-isocyanato-, ethyl ester, 3-isocyanatopropionic acid ethyl ester, ethyl n-oxomethylidene-beta-alaninate, ethyl n-oxomethylene-beta-alaninate, acmc-20akzk, ethyl-3-isocyanatopropionate, ethyl 3-isocyanato-propionate, ethyl 3-isocyanatopropion-ate, ethyl-3-isocyanato-propionate | |
XBSGYVHOINMTIM-UHFFFAOYSA-N | |
ethyl 3-isocyanatopropanoate | |
2733373 | |
98% |
Chemical Identifiers
5100-34-5 | |
143.14 | |
XBSGYVHOINMTIM-UHFFFAOYSA-N | |
2733373 | |
CCOC(=O)CCN=C=O |
C6H9NO3 | |
MFCD00078461 | |
ethyl 3-isocyanatopropionate, propanoic acid, 3-isocyanato-, ethyl ester, 3-isocyanatopropionic acid ethyl ester, ethyl n-oxomethylidene-beta-alaninate, ethyl n-oxomethylene-beta-alaninate, acmc-20akzk, ethyl-3-isocyanatopropionate, ethyl 3-isocyanato-propionate, ethyl 3-isocyanatopropion-ate, ethyl-3-isocyanato-propionate | |
ethyl 3-isocyanatopropanoate |
Safety and Handling
GHS H Statement
H331-H334-H227-H302-H315-H319-H335
Toxic if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Combustible liquid.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c
H302+H312+H332
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ISOCYANATES, TOXIC, N.O.S.
TSCA : No
Recommended Storage : Keep cold
RUO – Research Use Only