Learn More
Ethyl 3-isocyanatopropionate, 98%, Thermo Scientific Chemicals
$89.71 - $249.04
Chemical Identifiers
CAS | 5100-34-5 |
---|---|
Molecular Formula | C6H9NO3 |
Molecular Weight (g/mol) | 143.14 |
MDL Number | MFCD00078461 |
InChI Key | XBSGYVHOINMTIM-UHFFFAOYSA-N |
Synonym | ethyl 3-isocyanatopropionate, propanoic acid, 3-isocyanato-, ethyl ester, 3-isocyanatopropionic acid ethyl ester, ethyl n-oxomethylidene-beta-alaninate, ethyl n-oxomethylene-beta-alaninate, acmc-20akzk, ethyl-3-isocyanatopropionate, ethyl 3-isocyanato-propionate, ethyl 3-isocyanatopropion-ate, ethyl-3-isocyanato-propionate |
PubChem CID | 2733373 |
IUPAC Name | ethyl 3-isocyanatopropanoate |
SMILES | CCOC(=O)CCN=C=O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAL0961603
|
Thermo Scientific Chemicals
L0961603 |
1 g |
Each for $89.71
|
|
|||||
AAL0961606
|
Thermo Scientific Chemicals
L0961606 |
5 g |
Each for $249.04
|
|
|||||
Description
Ethyl 3-isocyanatopropionate can be used to produce 3-[1-(2-dimethylamino-ethylcarbamoyl)-5,6-dimethyl-6H-pyrido[4,3-b]carbazol-9-yloxycarbonylamino]-propionic acid ethyl ester with 9-hydroxy-5,6-dimethyl-6H-pyrido[4,3-b]carbazole-1-carboxylic acid (2-dimethylamino-ethyl)-amide.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsEthyl 3-isocyanatopropionate can be used to produce 3-[1-(2-dimethylamino-ethylcarbamoyl)-5,6-dimethyl-6H-pyrido[4,3-b]carbazol-9-yloxycarbonylamino]-propionic acid ethyl ester with 9-hydroxy-5,6-dimethyl-6H-pyrido[4,3-b]carbazole-1-carboxylic acid (2-dimethylamino-ethyl)-amide.
Solubility
Not miscible with water.
Notes
Store at 4°C. Moisture Sensitive. Avoid moisture, heat and direct sunlight. Incompatible materials are amines, alcohols and heat. Store away from moisture and oxidizing agents.
Chemical Identifiers
5100-34-5 | |
143.14 | |
XBSGYVHOINMTIM-UHFFFAOYSA-N | |
2733373 | |
CCOC(=O)CCN=C=O |
C6H9NO3 | |
MFCD00078461 | |
ethyl 3-isocyanatopropionate, propanoic acid, 3-isocyanato-, ethyl ester, 3-isocyanatopropionic acid ethyl ester, ethyl n-oxomethylidene-beta-alaninate, ethyl n-oxomethylene-beta-alaninate, acmc-20akzk, ethyl-3-isocyanatopropionate, ethyl 3-isocyanato-propionate, ethyl 3-isocyanatopropion-ate, ethyl-3-isocyanato-propionate | |
ethyl 3-isocyanatopropanoate |
Specifications
5100-34-5 | |
74°C to 75°C (4 mmHg) | |
Irritating | |
1.429 | |
1 g | |
1705618 | |
ethyl 3-isocyanatopropionate, propanoic acid, 3-isocyanato-, ethyl ester, 3-isocyanatopropionic acid ethyl ester, ethyl n-oxomethylidene-beta-alaninate, ethyl n-oxomethylene-beta-alaninate, acmc-20akzk, ethyl-3-isocyanatopropionate, ethyl 3-isocyanato-propionate, ethyl 3-isocyanatopropion-ate, ethyl-3-isocyanato-propionate | |
XBSGYVHOINMTIM-UHFFFAOYSA-N | |
ethyl 3-isocyanatopropanoate | |
2733373 | |
98% |
1.087 | |
92°C (197°F) | |
C6H9NO3 | |
MFCD00078461 | |
UN2206 | |
Moisture sensitive | |
Not miscible with water. | |
CCOC(=O)CCN=C=O | |
143.14 | |
143.14 | |
Ethyl 3-isocyanatopropionate |
Safety and Handling
GHS H Statement
H331-H334-H227-H302-H315-H319-H335
Toxic if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Combustible liquid.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c
H302+H312+H332
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ISOCYANATES, TOXIC, N.O.S.
TSCA : No
Recommended Storage : Keep cold
RUO – Research Use Only