missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethyl 3-phenylglycidate, 90%, mixture of cis and trans, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 332650050

 View more versions of this product

Catalog No. AC332650050


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Ethyl 3-phenylglycidate
121-39-1
1.1200g/mL
110°C
88% min. (GC)
C11H12O3
MFCD00005123
18,42
ethyl 3-phenylglycidate, ethyl phenylglycidate, oxiranecarboxylic acid, 3-phenyl-, ethyl ester, ethyl 2,3-epoxy-3-phenylpropionate, ethyl 3-phenyloxiranecarboxylate, glycidic acid, 3-phenyl-, ethyl ester, 2-ethyl-3-phenylglycidate, fema no. 2454, ccris 2623, ethyl3-phenylglycidate
GOMAKLPNAAZVCJ-UHFFFAOYSA-N
ethyl 3-phenyloxirane-2-carboxylate
8469
90%
90%
Colorless to Yellow
96.0°C (0.5 mmHg)
Authentic
Glass bottle
1.5165 to 1.5205
5 mL
1.12
Solubility in water: insoluble
CCOC(=O)C1C(O1)C2=CC=CC=C2
192.21
192.21
Liquid

Chemical Identifiers

121-39-1
192.21
GOMAKLPNAAZVCJ-UHFFFAOYSA-N
ethyl 3-phenyloxirane-2-carboxylate
C11H12O3
MFCD00005123
8469
CCOC(=O)C1C(O1)C2=CC=CC=C2
Videos
Safety and Handling

Safety and Handling

GHS H Statement
May cause an allergic skin reaction.

GHS P Statement
Wear protective gloves/protective clothing.
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation or rash occurs: Get medical advice/attention.

GHS Signal Word: Warning

EINECSNumber : 204-467-3

RTECSNumber : MB4970000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only