missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethyl 3-phenylglycidate, 90%, mixture of cis and trans, Thermo Scientific Chemicals

$63.00 - $84.64

Chemical Identifiers

CAS 121-39-1
Molecular Formula C11H12O3
Molecular Weight (g/mol) 192.21
MDL Number MFCD00005123
InChI Key GOMAKLPNAAZVCJ-UHFFFAOYSA-N
PubChem CID 8469
IUPAC Name ethyl 3-phenyloxirane-2-carboxylate
SMILES CCOC(=O)C1C(O1)C2=CC=CC=C2
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC332650050
SDS View Documents
Thermo Scientific Chemicals
332650050
5 mL Glass bottle
Each for $63.00
 
AC332650500
SDS View Documents
Thermo Scientific Chemicals
332650500
50 mL Glass bottle
Each for $84.64
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

121-39-1
192.21
GOMAKLPNAAZVCJ-UHFFFAOYSA-N
ethyl 3-phenyloxirane-2-carboxylate
C11H12O3
MFCD00005123
8469
CCOC(=O)C1C(O1)C2=CC=CC=C2

Specifications

121-39-1
1.1200g/mL
110°C
88% min. (GC)
C11H12O3
MFCD00005123
18,42
ethyl 3-phenylglycidate, ethyl phenylglycidate, oxiranecarboxylic acid, 3-phenyl-, ethyl ester, ethyl 2,3-epoxy-3-phenylpropionate, ethyl 3-phenyloxiranecarboxylate, glycidic acid, 3-phenyl-, ethyl ester, 2-ethyl-3-phenylglycidate, fema no. 2454, ccris 2623, ethyl3-phenylglycidate
GOMAKLPNAAZVCJ-UHFFFAOYSA-N
ethyl 3-phenyloxirane-2-carboxylate
8469
90%
Ethyl 3-phenylglycidate, 90%
Colorless to Yellow
96.0°C (0.5 mmHg)
Authentic
Glass bottle
1.5165 to 1.5205
5 mL
1.12
Solubility in water: insoluble
CCOC(=O)C1C(O1)C2=CC=CC=C2
192.21
192.21
Liquid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
May cause an allergic skin reaction.

GHS P Statement
Wear protective gloves/protective clothing.
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation or rash occurs: Get medical advice/attention.

GHS Signal Word: Warning

EINECSNumber : 204-467-3

RTECSNumber : MB4970000

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only