missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethyl 4,6-dichloronicotinate, 95%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals H6309206

 View more versions of this product

Catalog No. AAH6309206

Explore available promotions

Description

Description

Ethyl 4,6-dichloronicotinate, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff. It is also used as a fine chemical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl 4,6-dichloronicotinate, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff. It is also used as a fine chemical intermediate.

Solubility
Soluble in methanol.

Notes
Heat sensitive. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
Specifications

Specifications

Ethyl 4,6-dichloronicotinate
Yellow
MFCD00234408
ethyl 4,6-dichloronicotinate, 4,6-dichloronicotinic acid ethyl ester, 4,6-dichloropyridine-3-carboxylic acid ethyl ester, 3-pyridinecarboxylic acid, 4,6-dichloro-, ethyl ester, 2,4-dichloro-5-carbethoxypyridine, 4,6-dichloro-3-pyridinecarboxylic acid ethyl ester, 4,6-dichloro-nicotinic acid ethyl ester, 2,4-dichloro-5-ethoxycarbonyl pyridine, ethyl 2,4-dichloropyridine-5-carboxylate, pubchem17888
AAUBVINEXCCXOK-UHFFFAOYSA-N
ethyl 4,6-dichloropyridine-3-carboxylate
600045
95%
40296-46-6
C8H7Cl2NO2
5 g
Soluble in methanol.
CCOC(=O)C1=CN=C(C=C1Cl)Cl
220.049
220.05
Liquid

Chemical Identifiers

40296-46-6
220.049
AAUBVINEXCCXOK-UHFFFAOYSA-N
600045
CCOC(=O)C1=CN=C(C=C1Cl)Cl
C8H7Cl2NO2
MFCD00234408
ethyl 4,6-dichloronicotinate, 4,6-dichloronicotinic acid ethyl ester, 4,6-dichloropyridine-3-carboxylic acid ethyl ester, 3-pyridinecarboxylic acid, 4,6-dichloro-, ethyl ester, 2,4-dichloro-5-carbethoxypyridine, 4,6-dichloro-3-pyridinecarboxylic acid ethyl ester, 4,6-dichloro-nicotinic acid ethyl ester, 2,4-dichloro-5-ethoxycarbonyl pyridine, ethyl 2,4-dichloropyridine-5-carboxylate, pubchem17888
ethyl 4,6-dichloropyridine-3-carboxylate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280i-P304+P340-P305+P351+P338-P312-P501c

H319-H335

TSCA : No

storageNote1 : -30° to -10°C

Recommended Storage : Store at -20°C

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only