missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethyl 4,6-dichloronicotinate, 95%, Thermo Scientific Chemicals

$126.55 - $436.45

Chemical Identifiers

CAS 40296-46-6
Molecular Formula C8H7Cl2NO2
Molecular Weight (g/mol) 220.049
MDL Number MFCD00234408
InChI Key AAUBVINEXCCXOK-UHFFFAOYSA-N
Synonym ethyl 4,6-dichloronicotinate, 4,6-dichloronicotinic acid ethyl ester, 4,6-dichloropyridine-3-carboxylic acid ethyl ester, 3-pyridinecarboxylic acid, 4,6-dichloro-, ethyl ester, 2,4-dichloro-5-carbethoxypyridine, 4,6-dichloro-3-pyridinecarboxylic acid ethyl ester, 4,6-dichloro-nicotinic acid ethyl ester, 2,4-dichloro-5-ethoxycarbonyl pyridine, ethyl 2,4-dichloropyridine-5-carboxylate, pubchem17888
PubChem CID 600045
IUPAC Name ethyl 4,6-dichloropyridine-3-carboxylate
SMILES CCOC(=O)C1=CN=C(C=C1Cl)Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6309203
View Documents
Thermo Scientific Chemicals
H6309203
1 g
Each for $126.55
 
AAH6309206
View Documents
Thermo Scientific Chemicals
H6309206
5 g
Each for $436.45
 
Description

Description

Ethyl 4,6-dichloronicotinate, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff. It is also used as a fine chemical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl 4,6-dichloronicotinate, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff. It is also used as a fine chemical intermediate.

Solubility
Soluble in methanol.

Notes
Heat sensitive. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
Specifications

Chemical Identifiers

40296-46-6
220.049
AAUBVINEXCCXOK-UHFFFAOYSA-N
600045
CCOC(=O)C1=CN=C(C=C1Cl)Cl
C8H7Cl2NO2
MFCD00234408
ethyl 4,6-dichloronicotinate, 4,6-dichloronicotinic acid ethyl ester, 4,6-dichloropyridine-3-carboxylic acid ethyl ester, 3-pyridinecarboxylic acid, 4,6-dichloro-, ethyl ester, 2,4-dichloro-5-carbethoxypyridine, 4,6-dichloro-3-pyridinecarboxylic acid ethyl ester, 4,6-dichloro-nicotinic acid ethyl ester, 2,4-dichloro-5-ethoxycarbonyl pyridine, ethyl 2,4-dichloropyridine-5-carboxylate, pubchem17888
ethyl 4,6-dichloropyridine-3-carboxylate

Specifications

40296-46-6
C8H7Cl2NO2
1 g
Soluble in methanol.
CCOC(=O)C1=CN=C(C=C1Cl)Cl
220.049
220.05
Liquid
Yellow
MFCD00234408
ethyl 4,6-dichloronicotinate, 4,6-dichloronicotinic acid ethyl ester, 4,6-dichloropyridine-3-carboxylic acid ethyl ester, 3-pyridinecarboxylic acid, 4,6-dichloro-, ethyl ester, 2,4-dichloro-5-carbethoxypyridine, 4,6-dichloro-3-pyridinecarboxylic acid ethyl ester, 4,6-dichloro-nicotinic acid ethyl ester, 2,4-dichloro-5-ethoxycarbonyl pyridine, ethyl 2,4-dichloropyridine-5-carboxylate, pubchem17888
AAUBVINEXCCXOK-UHFFFAOYSA-N
ethyl 4,6-dichloropyridine-3-carboxylate
600045
95%
Ethyl 4,6-dichloronicotinate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280i-P304+P340-P305+P351+P338-P312-P501c

H319-H335

TSCA : No

storageNote1 : -30° to -10°C

Recommended Storage : Store at -20°C

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only